(-)-Dimethyl-D-tartrat zur Synthese

Lieferant: Merck

8.18723.0010
8.18723.0010EA 0 EUR
8.18723.0010
(-)-Dimethyl-D-tartrat zur Synthese
(-)-Dimethyl-D-tartrat
Formel: C₆H₁₀O₆
Siedepunkt: 245 °C (1013 hPa)
Schmelzpunkt: 45…48 °C
MDL: MFCD00008445
CAS-Nummer: 13171-64-7
EINECS: 236-118-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR