o-Ethyltoluol zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1-Ethyl-2-methylbenzol

M-Clarity™ Qualitätsstufe = MQ200

8.20044.0010
8.20044.0010EA 101 EUR
8.20044.0010
o-Ethyltoluol zur Synthese, Sigma-Aldrich®
o-Ethyltoluol
Formel: C₉H₁₂
Molecular Weight: 120,19 g/mol
Siedepunkt: 164 °C (1013 hPa)
Schmelzpunkt: –17 °C
Dichte: 0,88 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009257
CAS-Nummer: 611-14-3
EINECS: 210-255-1
UN: 3295
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Hill Formula C₉H₁₂
Chemical formula 2-(C₂H₅)C₆H₄CH₃
Molar Mass 120.19 g/mol
Boiling point 164 - 165 °C (1013 hPa)
Density 0.88 g/cm³ (20 °C)
Flash point 53 °C
Ignition temperature 440 °C
Melting Point -17 °C
Saturation concentration (air) 14 g/m³ (20 °C)
Vapor pressure 2.8 hPa (20 °C)
Refractive Index 1.5039 (20 °C, 589 nm)
Solubility 74.6 mg/l (25 °C) (experimental)
LD 50 oral LD50 Rat 5000 mg/kg
Storage class 3 Flammable liquids
WGK WGK 2 water endangering
R Phrase R 10
S Phrase S 24/25

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR