S-(2-Aminoethyl)isothiouronium bromide hydrobromide zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: S-(2-Aminoethyl)isothiourea Dihydrobromide, 2-(2-Aminoethyl)-2-thiopseudourea Dihydrobromide, S-(2-Aminoethyl)thiouronium bromide hydrobromide, AET, Carbamimidothioic Acid 2-Aminoethyl Ester Dihydrobromide

M-Clarity™ Qualitätsstufe = MQ200

8.20064.0010
8.20064.0010EA 64.3 EUR
8.20064.0010
S-(2-Aminoethyl)isothiouronium bromide hydrobromide zur Synthese, Sigma-Aldrich®
S-(2-Aminoethyl)isothiouronium bromide hydrobromide
Formel: C₃H₁₁Br₂N₃S
Molecular Weight: 281,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037011
CAS-Nummer: 56-10-0
EINECS: 200-257-0

Bestellen

Spezifikation Testergebnisse

Solubility 50 g/l (20 °C)
Melting Point 188 - 191 °C
Molar Mass 281.02 g/mol
Synonyms AET
Hill Formula C3H11Br2N3S
Chemical Formula H2NCH2CH2SC(=NH)NH2 * 2HBr

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR