Anthracen zur Synthese, Sigma-Aldrich®
Lieferant: Merck
8.20109.0100
8.20109.0250
8.20109.0100EA
44.7
EUR
8.20109.0100
8.20109.0250
Anthracen zur Synthese, Sigma-Aldrich®
Anthracen
Spezifikation Testergebnisse
Solubility |
|
Melting Point | 213 - 216 °C |
Molar Mass | 178.23 g/mol |
Bulk Density | 330 kg/m3 |
Boiling Point | 340 °C (1013 hPa) |
Vapor Pressure | 0.000008 hPa (25 °C) |
Flash Point | 121 °C |
Explosion Limit | 0.6 %(V) |
Ignition Point | 540 °C |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...