Benzaldehydoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: alpha-benzaldoxime, Benzaldoxime

M-Clarity™ Qualitätsstufe = MQ200

8.20234.0025
8.20234.0025EA 98.8 EUR
8.20234.0025
Benzaldehydoxim, Sigma-Aldrich®
Benzaldehydoxim
Formel: C₇H₇NO
Molecular Weight: 121,14 g/mol
MDL: MFCD00002119
CAS-Nummer: 932-90-1
EINECS: 213-261-2

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) slightly soluble
Melting Point 24 - 27 °C
Molar Mass 121.14 g/mol
Density 1.11 g/cm3
Synonyms Benzaldoxime
Hill Formula C7H7NO
Chemical Formula C6H5CH=NOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR