1,2,3,6-Tetrahydrophthalimid zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrahydrophthalimide

M-Clarity™ Qualitätsstufe = MQ200

8.20370.0250
8.20370.0250EA 63.5 EUR
8.20370.0250
1,2,3,6-Tetrahydrophthalimid zur Synthese, Sigma-Aldrich®
1,2,3,6-Tetrahydrophthalimid
Formel: C₈H₉NO₂
Molecular Weight: 151,1626 g/mol
MDL: MFCD00042782
CAS-Nummer: 85-40-5
EINECS: 201-602-8

Bestellen

Spezifikation Testergebnisse

Solubility 15 g/l (20 °C)
Melting Point 130 - 135 °C
Molar Mass 151.16 g/mol
Flash Point 178 °C
Synonyms 4-Cyclohexene-1,2-dicarboxylic acid imide, 4-Cyclohexene-1,2-dicarboximide
Hill Formula C8H9NO2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR