1,1-Dimethylharnstoff, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N-Dimethylharnstoff

M-Clarity™ Qualitätsstufe = MQ200

8.20509.0050
8.20509.0050EA 119 EUR
8.20509.0050
1,1-Dimethylharnstoff, Sigma-Aldrich®
1,1-Dimethylharnstoff
Formel: C₃H₈N₂O
Molecular Weight: 88,11 g/mol
MDL: MFCD00007959
CAS-Nummer: 598-94-7
EINECS: 209-957-0

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 180 - 183 °C
Molar Mass 88.11 g/mol
Synonyms 1,1-Dimethylurea
Hill Formula C3H8N2O
Chemical Formula (CH3)2NCONH2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR