2-Naphthylphenylketon zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Benzoylnaphthalene

M-Clarity™ Qualitätsstufe = MQ200

8.20869.0001
8.20869.0001EA 45.2 EUR
8.20869.0001
2-Naphthylphenylketon zur Synthese, Sigma-Aldrich®
2-Naphthyl phenyl ketone
Formel: C₁₇H₁₂O
Molecular Weight: 232,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004106
CAS-Nummer: 644-13-3
EINECS: 211-410-6

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Melting Point 80 - 82 °C
Molar Mass 232.28 g/mol
Boiling Point 398 °C
Synonyms Phenyl 2-naphthyl ketone
Hill Formula C17H12O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR