Anilin zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

8.22256.0100 8.22256.1000
8.22256.0100EA 22.6 EUR
8.22256.0100 8.22256.1000
Anilin zur Synthese, Sigma-Aldrich®
Anilin
Formel: C₆H₅NH₂
Siedepunkt: 184 °C (1013 hPa)
Dichte: 1,019 g/cm³ (20 °C)
Flammpunkt: 76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007629
CAS-Nummer: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II
Merck Index: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Hill Formula C₆H₇N
Chemical formula C₆H₅NH₂
Molar Mass 93.13 g/mol
Boiling point 184 °C (1013 hPa)
Density 1.02 g/cm³ (20 °C)
Explosion limit 1.2 - 11 %(V)
Flash point 76 °C
Ignition temperature 540 °C
Melting Point -6.2 °C
pH value 8.8 (36 g/l, H₂O, 20 °C)
Vapor pressure 0.5 hPa (20 °C)
Refractive Index 1.5863 (20 °C, 589 nm)
Solubility 36 g/l (20 °C)
LD 50 dermal LD50 Rabbit 840 mg/kg
Storage class 6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK WGK 2 water endangering
R Phrase R 23/24/25-40-41-43-48/23/24/25-68-50
S Phrase S 26-27-36/37/39-45-46-61-63


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR