Hydroxylammoniumchlorid zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Hydroxylamin-Hydrochlorid, Chlorhydrathydroxylamin, Hydroxylamin Hydrochlorid, Hydroxylamin HCl

M-Clarity™ Qualitätsstufe = MQ200

8.22334.9025
8.22334.9025EA 0 EUR
8.22334.9025
Hydroxylammoniumchlorid zur Synthese, Sigma-Aldrich®
Hydroxylammoniumchlorid
Formel: NH₂OH·HCl
Molecular Weight: 69,49 g/mol
Siedepunkt: 306 °C (1013 hPa)
Schmelzpunkt: 151 °C
Dichte: 1,67 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00051089
CAS-Nummer: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 (6.1),III
Merck Index: 13,04853

Bestellen

Spezifikation Testergebnisse

Grade Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Synonyms Hydroxylammonium Chloride
Chemical Formula NH2OH·HCl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR