(1R,2R)-(-)-1,2-Cyclohexandicarbonsäure zur Synthese
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.24449.0250
8.24449.0250EA
0
EUR
8.24449.0250
(1R,2R)-(-)-1,2-Cyclohexandicarbonsäure zur Synthese
(1R,2R)-(-)-1,2-Cyclohexandicarbonsäure
Formel:
C₈H₁₂O₄ |
MDL:
MFCD00062973 CAS-Nummer: 46022-05-3 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 177 - 180 °C |
Molar Mass | 172.18 g/mol |
Synonyms | (1R,2R)-(-)-Hexahydrophthalic acid |
Hill Formula | C8H12O4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...