(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride zur Synthese, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.24456.0500
8.24456.0500EA
72.3
EUR
8.24456.0500
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride zur Synthese, Sigma-Aldrich®
(2S,4R)-(-)-Ethyl-4-hydroxy-2-pyrrolidincarboxylate hydrochloride
Formel:
C₇H₁₃NO₃·HCl Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040589 CAS-Nummer: 33996-30-4 EINECS: 251-779-0 |
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Melting Point | 143 - 146 °C |
Molar Mass | 195.64 g/mol |
Synonyms | L-4-Hydroxyproline ethyl ester hydrochloride |
Hill Formula | C7H14ClNO3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...