Cyclohexadienyliumiron(0)tricarbonyl tetrafluoroborate zur Synthese
Lieferant: Merck
|
Gefahr
|
Synonyme:
CYCLOHEXADIENYLIUMIRON(0)-TRICARBONYL TETRAFLUOROBORATE
M-Clarity™ Qualitätsstufe = MQ200
8.24504.0250
8.24504.0250EA
0
EUR
8.24504.0250
Cyclohexadienyliumiron(0)tricarbonyl tetrafluoroborate zur Synthese
Cyclohexadienyliumiron(0)tricarbonyl tetrafluoroborate
Formel:
C₉H₇BF₄FeO₃ Molecular Weight: 305,8 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013293 CAS-Nummer: 33678-01-2 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Solubility | Soluble (20 °C) |
Melting Point |
|
Molar Mass | 305.8 g/mol |
Melting point |
|
Storage temperature | +15...+25 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...