Bis(4-fluorphenyl)methan zur Synthese
Lieferant: Merck
Synonyme:
4,4'-Difluorodiphenylmethane
M-Clarity™ Qualitätsstufe = MQ200
8.40056.0010
8.40056.0010EA
44
EUR
8.40056.0010
Bis(4-fluorphenyl)methan zur Synthese
Bis(4-fluorphenyl)methan
Formel:
C₁₃H₁₀F₂ Molecular Weight: 204,22 g/mol |
MDL:
MFCD00000360 CAS-Nummer: 457-68-1 EINECS: 207-274-2 |
Spezifikation Testergebnisse
Solubility | (20 °C) slightly soluble |
Molar Mass | 204.22 g/mol |
Boiling Point | 265 °C |
Density | 1.16 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...