N-Benzyl-N-methylanilin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.40106.0025
8.40106.0025EA
64.4
EUR
8.40106.0025
N-Benzyl-N-methylanilin, Sigma-Aldrich®
N-Benzyl-N-methylanilin
Formel:
C₁₄H₁₅N Dichte: 1,046 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035788 CAS-Nummer: 614-30-2 EINECS: 210-375-4 UN: 1602 ADR: 6.1,III |
Spezifikation Testergebnisse
Solubility | 0.009 g/l (20 °C) |
Melting Point | 8 °C |
Molar Mass | 197.28 g/mol |
Boiling Point | 311 °C (1013 hPa) |
Flash Point | 152 °C |
Density | 1.046 g/cm3 (20 °C) |
Ignition Point | 450 °C DIN 51794 |
Synonyms | Benzylmethylphenyl amine |
Hill Formula | C14H15N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...