4-(2-Pyridylazo)resorcin zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(2'-Pyridylazo)resorcinol

M-Clarity™ Qualitätsstufe = MQ200

8.41016.0005
8.41016.0005EA 244 EUR
8.41016.0005
4-(2-Pyridylazo)resorcin zur Synthese, Sigma-Aldrich®
4-(2-Pyridylazo)resorcin
Formel: C₁₁H₉N₃O₂
Molecular Weight: 215,21 g/mol
MDL: MFCD00044237
CAS-Nummer: 1141-59-9
EINECS: 214-528-6

Bestellen

Spezifikation Testergebnisse

Melting Point 187 - 189 °C
Molar Mass 215.21 g/mol
Synonyms 4-(2-Pyridinylazo)-1,3-dihydroxybenzene
Hill Formula C11H9N3O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR