Aminoacetonitril-Sulfat zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Aminoacetonitrile hemisulphate, Glycinonitrile sulphate, Cyanomethylamine sulphate

M-Clarity™ Qualitätsstufe = MQ200

8.41034.0050
8.41034.0050EA 138 EUR
8.41034.0050
Aminoacetonitril-Sulfat zur Synthese, Sigma-Aldrich®
Aminoacetonitrile sulphate
Formel: C₄H₁₀N₄O₄S
Molecular Weight: 210,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012851
CAS-Nummer: 5466-22-8
EINECS: 226-772-0
UN: 3439
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Melting Point 118 - 122 °C
Molar Mass 210.2 g/mol
Synonyms Glycinonitrile sulphate
Hill Formula C4H10N4O4S
Storage temperature +15...+25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR