2'-Chloracetophenon zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: o-Chloracetophenon, 1-(2-Chlorphenyl)ethanon

M-Clarity™ Qualitätsstufe = MQ200

8.41072.0050
8.41072.0050EA 116 EUR
8.41072.0050
2'-Chloracetophenon zur Synthese, Sigma-Aldrich®
2'-Chloracetophenon
Formel: C₈H₇ClO
Molecular Weight: 154,6 g/mol
Siedepunkt: 227 °C (1013 hPa)
Dichte: 1,189 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000560
CAS-Nummer: 2142-68-9
EINECS: 218-397-6
UN: 3416
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Hill Formula C₈H₇ClO
Molar Mass 154.59 g/mol
Storage class 6.1B Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK WGK 3 highly water endangering
R Phrase R 22-36/37/38

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR