Ethyl-4-(dimethylamino)benzoat zur Synthese, Sigma-Aldrich®
Lieferant: Merck
8.41086.0100
8.41086.9999
8.41086.0100EA
117
EUR
8.41086.0100
8.41086.9999
Ethyl-4-(dimethylamino)benzoat zur Synthese, Sigma-Aldrich®
Ethyl-4-(dimethylamino)benzoat
Spezifikation Testergebnisse
Melting Point | 63 - 65 °C |
Molar Mass | 193.24 g/mol |
Synonyms | 4-Dimethylaminobenzoic acid ethyl ester |
Hill Formula | C11H15NO2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...