N-tert-Butyl-α-Phenylnitron zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
PBN, N-Benzylidene-tert-butylamine N-Oxide, N-tert-Butyl-α-phenylnitrone
M-Clarity™ Qualitätsstufe = MQ200
8.41179.0001
8.41179.0001EA
122
EUR
8.41179.0001
N-tert-Butyl-α-Phenylnitron zur Synthese, Sigma-Aldrich®
N-tert-Butyl-α-Phenylnitron
Formel:
C₁₁H₁₅NO Lagertemperatur: Kühlschrank |
MDL:
MFCD00008799 CAS-Nummer: 3376-24-7 EINECS: 222-168-6 |
Spezifikation Testergebnisse
Solubility | Insoluble at 20 °C |
Melting Point | 71...73 °C |
Molar Mass | 177,24 g/mol |
HS Code | 2929 90 00 |
Storage temperature | Store below +15 °C |
Assay (HPLC, area%) | ≥98% (a/a) |
Melting range (lower value) | ≥71 °C |
Melting range (upper value) | ≤73 °C |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...