Propargyltriphenylphosphoniumbromid zur Synthese
Lieferant: Merck
|
Achtung
|
Synonyme:
2-Propinyl-triphenylphosphonium bromide, Triphenyl(prop-2-yn-1-yl)phosphonium bromide
8.41338.0005
8.41338.0005EA
112
EUR
8.41338.0005
Propargyltriphenylphosphoniumbromid zur Synthese
Propargyltriphenylphosphoniumbromid
Formel:
C₂₁H₁₈BrP Molecular Weight: 381,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011811 CAS-Nummer: 2091-46-5 EINECS: 218-242-2 UN: 3464 ADR: 6.1,III |
Spezifikation Testergebnisse
Melting Point | 177 - 178 °C |
Molar Mass | 381.25 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...