Ethyl(trimethylsilyl)acetat

Lieferant: Merck

8.41404.0005
8.41404.0005EA 0 EUR
8.41404.0005
Ethyl(trimethylsilyl)acetat
Ethyl(trimethylsilyl)acetat
Formel: C₇H₁₆O₂Si
MDL: MFCD00009172
CAS-Nummer: 4071-88-9
EINECS: 223-783-2
UN: 3272
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR