3-Fluor-DL-phenylalanin zur Synthese

Lieferant: Merck

8.41409.0005
8.41409.0005EA 0 EUR
8.41409.0005
3-Fluor-DL-phenylalanin zur Synthese
3-Fluor-DL-phenylalanin
Formel: C₉H₁₀FNO₂
MDL: MFCD00004273
CAS-Nummer: 2629-54-1
EINECS: 220-104-1
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR