6-Methoxysalicylaldehyd

Lieferant: Merck

Synonyme: 2-Hydroxy-6-methoxybenzaldehyde

8.41484.0001
8.41484.0001EA 0 EUR
8.41484.0001
6-Methoxysalicylaldehyd
6-Methoxysalicylaldehyd
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00151830
CAS-Nummer: 700-44-7
EINECS: 211-844-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR