β-Cholestanol zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
beta-Cholestanol, 5A-Cholestan-3B-Ol, 5-ɑ-Cholestan-3-β-ol, Dihydrocholesterol
M-Clarity™ Qualitätsstufe = MQ200
8.41513.0005
8.41513.0005EA
114
EUR
8.41513.0005
β-Cholestanol zur Synthese, Sigma-Aldrich®
beta-Cholestanol
Formel:
C₂₇H₄₈O Molecular Weight: 388,68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066413 CAS-Nummer: 80-97-7 EINECS: 201-315-8 |
Spezifikation Testergebnisse
Melting Point | 130...143 °C |
Molar Mass | 388,67 g/mol |
EC number | 201-315-8 |
Hill Formula | C27H48O |
HS Code | 2906 13 10 |
Storage temperature | 15...25 °C |
Assay (GC, area%) | ≥98% (a/a) |
Melting range (lower value) | ≥139 °C |
Melting range (upper value) | ≤143 °C |
Spec. rotation α²0/D (c=1 in chloroform) | 22,0 - 25,0 ° |
Identity (IR) | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...