Benzo[b]thiophen, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Thianaphthene, 1-Benzothiophene
M-Clarity™ Qualitätsstufe = MQ200
8.41538.0010
8.41538.0010EA
54.7
EUR
8.41538.0010
Benzo[b]thiophen, Sigma-Aldrich®
Benzo[b]thiophen
Formel:
C₈H₆S Molecular Weight: 134.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005864 CAS-Nummer: 95-15-8 EINECS: 202-395-7 UN: 3077 ADR: 9 III |
Spezifikation Testergebnisse
Solubility | 0.13 g/l (25 °C) |
Melting Point | 28 - 32 °C |
Molar Mass | 134.19 g/mol |
Boiling Point | 222 °C |
Vapor Pressure | 1.33 hPa (20 °C) |
Flash Point | 91 °C |
Density | 1.15 g/cm3 |
Ignition Point | 510 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...