Benzotrifluorid zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
α,α,α-Trifluortoluol, Lösungsmittel BTF
M-Clarity™ Qualitätsstufe = MQ200
8.41554.0100
8.41554.0100EA
51.7
EUR
8.41554.0100
Benzotrifluorid zur Synthese, Sigma-Aldrich®
Benzotrifluorid
Formel:
C₇H₅F₃ Molecular Weight: 146,11 g/mol Siedepunkt: 101 °C (1013 hPa) Schmelzpunkt: –29 °C Dichte: 1,37 g/cm³ (20 °C) Flammpunkt: 12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000372 CAS-Nummer: 98-08-8 EINECS: 202-635-0 UN: 2338 ADR: 3,II Merck Index: 13,01111 |
Spezifikation Testergebnisse
Hill Formula | C₇H₅F₃ |
Molar Mass | 146.11 g/mol |
Boiling point | 100 - 103 °C (1013 hPa) |
Density | 1.197 g/cm³ (20 °C) |
Explosion limit | 1.4 - 9.3 %(V) |
Flash point | 12 °C |
Ignition temperature | 650 °C |
Melting Point | -29 °C |
Vapor pressure | 53 hPa (25 °C) |
Solubility | 0.45 g/l (20 °C) Hydrolysis |
LD 50 oral | LD50 Rat 15000 mg/kg |
LD 50 dermal |
LD50 Rat |
Storage class | 3 Flammable liquids |
WGK | WGK 1 slightly water endangering |
R Phrase | R 11-51/53 |
S Phrase | S 16-23-61 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...