4,4'-Anisoin zur Synthese
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.41595.0025
8.41595.0025EA
59
EUR
8.41595.0025
4,4'-Anisoin zur Synthese
4,4'-Anisoin
Formel:
C₁₆H₁₆O₄ |
MDL:
MFCD00008411 CAS-Nummer: 119-52-8 EINECS: 204-330-8 |
Spezifikation Testergebnisse
Melting Point | 111 - 112 °C |
Molar Mass | 272.3 g/mol |
Synonyms | 4,4'-Dimethoxybenzoin |
Hill Formula | C16H16O4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...