Diphenylborsäure(2-aminoethyl)ester (2-APB) ≥98,0% zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine

M-Clarity™ Qualitätsstufe = MQ200

8.41636.0005 8.41636.8500
8.41636.0005EA 91.1 EUR
8.41636.0005 8.41636.8500
Diphenylborsäure(2-aminoethyl)ester (2-APB) ≥98,0% zur Synthese, Sigma-Aldrich®
Diphenylborsäure(2-aminoethyl)ester (2-APB)
Formel: C₁₄H₁₆BNO
Molecular Weight: 225.1 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00014823
CAS-Nummer: 524-95-8
EINECS: 208-366-5

Bestellen

Spezifikation Testergebnisse

CAS 524-95-8
Synonyms Diphenylboric acid-2-aminoethyl ester
Hill Formula C₁₄H₁₆BNO
Molar Mass 225.09 g/mol
Melting Point 189 - 194 °C
Storage Store below +15°C.
Assay (HClO₄) ≥ 98,0 %(m)
Identity (IR) conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR