Diphenylborsäure(2-aminoethyl)ester (2-APB) ≥98,0% zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2-Aminoethyl diphenylborinate, 2-((Diphenylboryl)oxy)ethanamine
M-Clarity™ Qualitätsstufe = MQ200
8.41636.0005
8.41636.8500
8.41636.0005EA
91.1
EUR
8.41636.0005
8.41636.8500
Diphenylborsäure(2-aminoethyl)ester (2-APB) ≥98,0% zur Synthese, Sigma-Aldrich®
Diphenylborsäure(2-aminoethyl)ester (2-APB)
Formel:
C₁₄H₁₆BNO Molecular Weight: 225.1 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00014823 CAS-Nummer: 524-95-8 EINECS: 208-366-5 |
Spezifikation Testergebnisse
CAS | 524-95-8 |
Synonyms | Diphenylboric acid-2-aminoethyl ester |
Hill Formula | C₁₄H₁₆BNO |
Molar Mass | 225.09 g/mol |
Melting Point | 189 - 194 °C |
Storage | Store below +15°C. |
Assay (HClO₄) | ≥ 98,0 %(m) |
Identity (IR) | conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...