4-(4-Nitrophenylazo)resorcin zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-((4-Nitrophenyl)diazenyl)benzene-1,3-diol, Azoviolet, Azo Violet, Magneson I
M-Clarity™ Qualitätsstufe = MQ200
8.41713.0025
8.41713.0025EA
124
EUR
8.41713.0025
4-(4-Nitrophenylazo)resorcin zur Synthese, Sigma-Aldrich®
4-(4-Nitrophenylazo)resorcin
Formel:
C₁₂H₉N₃O₄ Molecular Weight: 259,22 g/mol |
MDL:
MFCD00007310 CAS-Nummer: 74-39-5 EINECS: 200-808-5 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Melting Point | 183 °C (decomposition) |
Molar Mass | 259.22 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...