Cholesteryl chloride zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cholesteryl Chloride, (3S,8S,9S,10R,13R,14S,17R)-3-Chloro-10,13-dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
M-Clarity™ Qualitätsstufe = MQ200
8.41733.0010
8.41733.0010EA
63
EUR
8.41733.0010
Cholesteryl chloride zur Synthese, Sigma-Aldrich®
Cholesteryl chloride
Formel:
C₂₇H₄₅Cl Molecular Weight: 405.107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003632 CAS-Nummer: 910-31-6 EINECS: 213-004-4 |
Spezifikation Testergebnisse
Solubility | 542 g/l (20 °C) |
Melting Point | 712 °C |
Molar Mass | 95.22 g/mol |
Boiling Point | 1412 °C (1013 hPa) |
Density | 2.32 g/cm3 (20 °C) |
pH |
|
Synonyms | Magnesium dichloride |
Hill Formula | Cl2Mg |
Chemical Formula | MgCl2 |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...