3-Aminopropionitril-fumarat

Lieferant: Merck

8.41773.0025
8.41773.0025EA 0 EUR
8.41773.0025
3-Aminopropionitril-fumarat
3-Aminopropionitril-fumarat
Formel: C₁₀H₁₆N₄O₄
Molecular Weight: 256,26 g/mol
MDL: MFCD00013142
CAS-Nummer: 2079-89-2
EINECS: 218-208-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR