1,4-Bis(2-methylstyryl)benzene zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis-MSB, 1,4-Di(2-methylstyryl)benzene, 1-Methyl-2-(2-(4-[2-(2-methylphenyl)ethenyl]phenyl)ethenyl)benzene, p-Bis(o-Methylstyryl) Benzene

M-Clarity™ Qualitätsstufe = MQ200

8.41875.0001
8.41875.0001EA 41.6 EUR
8.41875.0001
1,4-Bis(2-methylstyryl)benzene zur Synthese, Sigma-Aldrich®
1,4-Bis(2-methylstyryl)benzene
Formel: C₂₄H₂₂
Molecular Weight: 310,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008529
CAS-Nummer: 13280-61-0
EINECS: 236-285-5

Bestellen

Spezifikation Testergebnisse

Melting Point 179...181 °C
Molar Mass 310.43 g/mol
Synonyms Bis-MSB
Hill Formula C24H22
HS Code 2902 90 00
Storage temperature 15...25 °C
Assay (HPLC, area%) ≥97% (a/a)
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR