3-Methylflavone-8-carboxylic acid ≥98% zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3-Methyl-2-phenylchromone-8-carboxylic Acid, 3-Methyl-4-oxo-2-phenyl-4H-chromene-8-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ200

8.41878.0005
8.41878.0005EA 91.7 EUR
8.41878.0005
3-Methylflavone-8-carboxylic acid ≥98% zur Synthese, Sigma-Aldrich®
3-Methylflavone-8-carboxylic acid
Formel: C₁₇H₁₂O₄
Molecular Weight: 280.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040998
CAS-Nummer: 3468-01-7
EINECS: 222-425-2

Bestellen

Spezifikation Testergebnisse

Melting Point 235...240 °C
Molar Mass 280,28 g/mol
Hill Formula C17H12O4
HS Code 2932 99 00
Storage temperature 15...25 °C
Assay (HPLC, area%) ≥98% (a/a)
Melting range (lower value) ≥235 °C
Melting range (upper value) ≤240 °C
Identity (IR) passes test

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR