2-(4-Chlorphenylthio)benzaldehyd zur Synthese

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

8.41881.0001
8.41881.0001EA 0 EUR
8.41881.0001
2-(4-Chlorphenylthio)benzaldehyd zur Synthese
2-(4-Chlorophenylthio)benzaldehyde
Formel: C₁₃H₉ClOS
MDL: MFCD00051766
CAS-Nummer: 107572-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR