Phenyltrimethoxysilan zur Synthese, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
8.43658.0050
8.43658.0050EA
89.4
EUR
8.43658.0050
Phenyltrimethoxysilan zur Synthese, Sigma-Aldrich®
Phenyltrimethoxysilan
Formel:
C₉H₁₄O₃Si Dichte: 1,064 g/cm³ (20 °C) |
MDL:
MFCD00025689 CAS-Nummer: 2996-92-1 EINECS: 221-066-9 |
Spezifikation Testergebnisse
Molar Mass | 198.3 g/mol |
Boiling Point | 233 °C |
Flash Point | 83 °C |
Density | 1.064 g/cm3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...