Diethylenglycolmethylethylether zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Ethoxy-2-(2-methoxyethoxy)ethane, Diethylene glycol ethyl methyll ether, 2,5,8-Trioxadecane
M-Clarity™ Qualitätsstufe = MQ200
8.43827.1000
8.43827.1000EA
132
EUR
8.43827.1000
Diethylenglycolmethylethylether zur Synthese, Sigma-Aldrich®
Diethylenglycolmethylethylether
Formel:
C₇H₁₆O₃ Molecular Weight: 148,2001 g/mol |
MDL:
MFCD01727263 CAS-Nummer: 1002-67-1 EINECS: 213-690-5 |
Spezifikation Testergebnisse
Solubility | 76 g/l (25 °C) |
Melting Point | -72 °C |
Molar Mass | 148.2 g/mol |
Flash Point | 82 °C |
Density | 0.925 g/cm3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...