3-Phenyl-1H-pyrazol zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole
M-Clarity™ Qualitätsstufe = MQ200
8.43855.0005
8.43855.0005EA
97.5
EUR
8.43855.0005
3-Phenyl-1H-pyrazol zur Synthese, Sigma-Aldrich®
3-Phenyl-1H-pyrazole
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159654 CAS-Nummer: 2458-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...