Benzethoniumchlorid

Lieferant: Merck
Gefahr

Synonyme: Hyamine® 1622

M-Clarity™ Qualitätsstufe = MQ200

8.43983.0250 8.43983.9999
8.43983.0250EA 302 EUR
8.43983.0250 8.43983.9999
Benzethoniumchlorid
Benzethoniumchlorid
Formel: C₂₇H₄₂ClNO₂
Schmelzpunkt: 164…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011742
CAS-Nummer: 121-54-0
EINECS: 204-479-9
UN: 2923
ADR: 8 (6.1),III
Merck Index: 13,01074

Bestellen

Spezifikation Testergebnisse

Hill Formula C₂₇H₄₂ClNO₂
Molar Mass 448.08 g/mol
Ignition temperature 380 °C
Melting Point 160 - 166 °C
Vapor pressure  - 0.001 hPa (25 °C)
Bulk density 440 kg/m³
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 295 mg/kg
Signal Word Danger
Storage class 6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK WGK 2 water endangering
R Phrase R 22-34-50/53
S Phrase S 26-36/37/39-45-61


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR