HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Novabiochem®
Lieferant: NOVABIOCHEM
|
Gefahr
|
8.51013.0025
8.51013.0100
8.51013.0500
8.51013.1000
01-62-0041-25EA
452
EUR
01-62-0041-25
8.51013.0100
NOVA8.51013.0500
NOVA8.51013.1000
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Novabiochem®
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Formel:
C₁₀H₁₅F₆N₆OP Schmelzpunkt: 186 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00274639 CAS-Nummer: 148893-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...