Immobiline™ DryPlate for IEF
Lieferant: Cytiva
80-1128-28
80-1128-28EA 965 EUR
80-1128-28
Immobiline™ DryPlate for IEF
Elektrophorese-Gele Fertiggele
Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients.
  • Choose from narrow (1 pH unit) and medium (3 pH units) pH ranges
  • Handle stable, plastic-backed gels with confidence
Immobiline™ DryPlate precast gels contain preformed, immobilized, non-drifting pH gradients. The gels are rehydrated to a thickness of 0.5 mm in the reswelling cassette accessory before use.
Immobiline™ technology produces a stable pH gradient, allowing high voltages and resolution over narrow pH ranges, even for low mobility proteins that require long focusing times.
Bestellinformation: Immobiline™ sample application strips include 52 samples, 5–20µL and instructions.
Lieferumfang: Kit includes 50× filter paper, 3× experimental result form instructions.