p-Xylene-bis(N-pyridinium bromide) (DPX)

Lieferant: Biotium
Achtung

Synonyme: 1,1'-(p-Xylylene)bis(1-pyridinium) dibromide, 1,1'-(1,4-Phenylenebis(methylene))bis(pyridin-1-ium) bromide, DPX, 1,1'-(p-Xylylene)bis(1-pyridinium bromide), 1,1'-[1,4-Phenylenebis(methylene)]bis(1-pyridinium) dibromide

80012
80012.EA 118 EUR
80012.
p-Xylene-bis(N-pyridinium bromide) (DPX)
p-Xylene-bis(N-pyridinium bromide) (DPX)

DPX is a positively charged quencher that is often used with ANTS (90010) to study membrane fusion or permeability.


  • Off-white solid soluble in water
  • Store desiccated at 4°C

Formel: C₁₈H₁₈Br₂N₂
Molecular Weight: 422,16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00043749
CAS-Nummer: 14208-10-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=746526d13034a255b48f8e54e2f92072eaf153e2&vid=746526d13034a255b48f8e54e2f92072eaf153e2&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR