Calcein

Lieferant: Biotium

80013
80013.EA 126 EUR
80013.
Calcein
Calcein
Formel: C₃₀H₂₆N₂O₁₃
MDL: MFCD00005049
CAS-Nummer: 1461-15-0
EINECS: 215-957-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR