Hydroxystilbamidine bis(methanesulphonate) fluoreszierender Farbstoff, für die Histologie

Lieferant: Biotium
Achtung

Synonyme: Fluoro-Gold™

80014
80014.EA 387 EUR
80014.
Hydroxystilbamidine bis(methanesulphonate) fluoreszierender Farbstoff, für die Histologie
Hydroxystilbamidine bis(methanesulphonate)

Hydroxystilbamidine (also called Fluoro-Gold™) has been used extensively as a retrograde tracer for neurons and also a histochemical stain. Fluoro-Gold™ can undergo retrograde axonal transport, it shows extensive filling of dendrites, and has a high resistance to fading.


  • Cellular localization: Fluid phase tracer
  • λEx/λEm = 361/536 nm
  • Yellow solid soluble in water
  • Store at 4°C and protect from light


Also see hydroxystilbamidine 4% in H2O (80023), which is a more convenient, ready-to-use form.

Formel: C₁₈H₂₄N₄O₇S₂
Molecular Weight: 472,54 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01863083
CAS-Nummer: 223769-64-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15b7e976e027d34cff19632022fb4557a19c2c23&vid=15b7e976e027d34cff19632022fb4557a19c2c23&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR