Lucifer yellow CH dipotassium salt fluoreszierender Farbstoff, für die Histologie
Lieferant: Biotium
Synonyme:
LY CH potassium salt
80016
80016.EA
139
EUR
80016.
Lucifer yellow CH dipotassium salt fluoreszierender Farbstoff, für die Histologie
Lucifer yellow CH dipotassium salt
Lucifer Yellow CH potassium salt (LY CH potassium salt) is a widely used polar molecular tracer for studying neuronal morphology. The fluorescent molecule contains a carbohydrazide that allow the molecule to be aldehyde-fixable.
- λEx/λ Em = 428/536 nm
- Yellow solid soluble in water
- Store at 4°C and protect from light
Formel:
C₁₃H₉K₂N₅O₉S₂ Molecular Weight: 521,57 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00013445 CAS-Nummer: 71206-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...