Lucifer Yellow Cadaverine Biotin-X dipotassium salt fluoreszierender Farbstoff, für die Histologie
Lieferant: Biotium
80017
80017.EA 257 EUR
80017.
Lucifer Yellow Cadaverine Biotin-X dipotassium salt fluoreszierender Farbstoff, für die Histologie
Lucifer Yellow Cadaverine Biotin-X dipotassium salt
This fluorescent biotin derivative is a useful tracer for studying neuronal morphology. The biotin portion of the molecule allows researchers to amplify the fluorescent signal using standard biotin-avidin or biotin-streptavidin biochemistry. The staining can tolerate aldehyde fixation, sectioning, and washing with detergent.
  • λAbs/λEms 428/532 nm
  • Yellow solid soluble in water
  • Store desiccated at ≤4°C and protect from light