Fluorescein biotin fluoreszierender Farbstoff, für die Histologie

Lieferant: Biotium

80019
80019.EA 217 EUR
80019.
Fluorescein biotin fluoreszierender Farbstoff, für die Histologie
Fluorescein biotin

Fluorescein biotin (5-((N-(5-(N-(6-(Biotinoyl)amino)hexanoyl)amino)pentyl)thioureidyl) fluorescein) can be used to detect and quantify biotin binding sites and the degree of biotinylation of proteins.


  • λEx/λ Em= 494/518 nm
  • Yellow solid soluble in DMSO, or DMF
  • Store at -20°C and protect from light


Also see biotin-rhodamine 110 (80022) and biotin-4-fluorescein (90062).

Formel: C₄₂H₅₀N₆O₈S₂
Lagertemperatur: Gefrierschrank
CAS-Nummer: 134759-22-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e4f9971ae5ceb51b98c74c0bdc3503ecbd567ea&vid=0e4f9971ae5ceb51b98c74c0bdc3503ecbd567ea&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR