Tris(bathophenanthrolinedisulphonate)ruthenium(II) sodium salt fluoreszierender Farbstoff, für die Histologie

Lieferant: Biotium

Synonyme: RuBPS

80021
80021.EA 319 EUR
80021.
Tris(bathophenanthrolinedisulphonate)ruthenium(II) sodium salt fluoreszierender Farbstoff, für die Histologie
Tris(bathophenanthrolinedisulphonate)ruthenium(II) sodium salt

RuBPS (Ruthenium(tris(bathophenanthrolinedisulfonate), tetrasodium salt) is a negatively-charged water-soluble luminescent ruthenium complex. The complex can be efficiently excited by either UV light or visible light in the wavelength range from 400 nm to 500 nm with bright red emission centered around 605 nm.


  • λEx/λEm(H2O) = 462 nm (broad)/603 nm
  • Yellow solid soluble in water
  • Store at 4°C and protect from light


Ruthenium complexes such as RuBPS typically have luminescence lifetimes in the micro second range and a Stokes shift over 100 nm. By using time-resolved luminescence measurement, the combination of long luminescent lifetime and large Stokes shift offers the possibility for extremely sensitive detection that could approach the sensitivity of radioactive detection. Among many potential applications, RuBPS could be used as a polar tracer similar to Lucifer Yellow and FluoroGold.

Formel: C₇₂H₄₂N₆Na₄O₁₈RuS₆
Molecular Weight: 1664,55 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04040034
CAS-Nummer: 301206-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dda8e3ddc98b2ef235f2e4637e214018e6b562d&vid=7dda8e3ddc98b2ef235f2e4637e214018e6b562d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR