RuBP-2S fluoreszierender Farbstoff, für die Histologie

Lieferant: Biotium

Synonyme: Ruthenium(bis(bathophenanthroline))(bathophenanthrolinedisulphonate

80024
80024.EA 340 EUR
80024.
RuBP-2S fluoreszierender Farbstoff, für die Histologie
RuBP-2S

RuBP-2S (Ruthenium(bis(bathophenanthroline))(bathophenanthrolinedisulfonate) is a luminescent ruthenium complex similar to RuBPS (80021). Two of the bathophenanthroline ligands are neutral while the third bathophenanthroline ligand is bis-sulfonated, resulting in an overall neutral charge of the ruthenium complex.


  • λEx/λEm(H2O)= 462 (broad)/603 nm
  • Yellow solid soluble in water
  • Store at 4°C and protect from light


RuBP-2S shares the spectral properties of RuBPS and may also have similar applications.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea19fc2f88b31fb8a8f9b899d9d0f43d55aba45d&vid=f869743c3eda9a2e6c36dc5ada610040c6bfc592&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR